accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2N

CAS RN 14328-51-9

IUPAC Name: 2-amino-2-cyclohexylacetic acid
Synonyms: cyclohexylglycine
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: WAMWSIDTKSNDCU-UHFFFAOYNA-N
SMILES: NC(C1CCCCC1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)