Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-2-Cyclohexylglycine, 98%

Catalog No. AAH2688106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2688106 5 g
AAH2688103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2688106 Supplier Thermo Scientific Chemicals Supplier No. H2688106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14328-51-9 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14328-51-9
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD01311679
InChI Key WAMWSIDTKSNDCU-ZETCQYMHSA-N
Synonym l-alpha-cyclohexylglycine, h-chg-oh, l-cyclohexylglycine, s-2-amino-2-cyclohexylacetic acid, h-cyclohexyl-gly-oh, l-+-2-cyclohexylglycine, 2s-amino cyclohexyl acetic acid, cyclohexylglycyl, 2-cyclohexyl-l-glycine, 2s-2-amino-2-cyclohexylacetic acid
PubChem CID 736848
IUPAC Name (2S)-2-amino-2-cyclohexylacetic acid
SMILES C1CCC(CC1)C(C(=O)O)N

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 157.22
Optical Rotation +31° (c=0.5 in 1N HCl)
Percent Purity 98%
Chemical Name or Material L-(+)-2-Cyclohexylglycine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.