accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOO

CAS RN 143536-99-6

IUPAC Name: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxane
Synonyms: 3,4,5-tris(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxane
Molecular Weight (g/mol): 646.78
Molecular Formula: C41H42O7
InChi Key: QXYYOIDTAHLOQE-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C2OCC2=CC=CC=C2)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results