Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenyl 2,3,4,6-Tetra-O-benzyl-beta-D-galactopyranoside 98.0+%, TCI America™
SDP

Catalog No. M15885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M15885G Supplier TCI America Supplier No. M15885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protected monosaccharide building block for glycosylation with 98.0% minimum purity.

Chemical Identifiers

CAS 143536-99-6
Molecular Formula C41H42O7
Molecular Weight (g/mol) 646.78
MDL Number MFCD06797131
InChI Key QXYYOIDTAHLOQE-WQLDJJBGSA-N
PubChem CID 11422359
IUPAC Name (2S,3R,4S,5S,6R)-2-(4-methoxyphenoxy)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxane
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5)OCC6=CC=CC=C6

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Formula Weight 646.78
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 2,3,4,6-Tetra-O-benzyl-beta-D-galactopyranoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.