accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14375-45-2

IUPAC Name: (2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
Synonyms: acide abscissique acido abscisico abszisinsaeure abscisinsaeure 2-cis-abscisic acid
Molecular Weight (g/mol): 264.32
Molecular Formula: C15H20O4
InChi Key: JLIDBLDQVAYHNE-LXGGSRJLNA-N
SMILES: C\C(\C=C\C1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)