Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Abscisic acid, 98%, Thermo Scientific™

Catalog No. AC133482500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133482500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC133482500 Supplier Thermo Scientific Chemicals Supplier No. 133482500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14375-45-2
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.321
InChI Key JLIDBLDQVAYHNE-LXGGSRJLSA-N
Synonym abscisic acid, +/--abscisic acid, acide abscissique, 2z,4e-5-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl-3-methylpenta-2,4-dienoic acid, abscisinsaeure, abszisinsaeure, acido abscisico, +-abscisic acid, 2-cis-abscisic acid, rs-abscisic acid
PubChem CID 5375199
ChEBI CHEBI:22152
IUPAC Name (2Z,4E)-5-(1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl)-3-methylpenta-2,4-dienoic acid
SMILES CC1=CC(=O)CC(C1(C=CC(=CC(=O)O)C)O)(C)C

Specifications

Melting Point 188.0°C to 191.0°C
Quantity 250 mg
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone, methanol, ethanol, chloroform,, diethyl ether, sodium hydroxide solution
Formula Weight 264.32
Chemical Name or Material (±)-Abscisic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.