accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 1438-94-4

IUPAC Name: 1-[(furan-2-yl)methyl]-1H-pyrrole
Synonyms: 1-furfuryl-1H-pyrrole
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: BTBFUBUCCJKJOZ-UHFFFAOYSA-N
SMILES: C(N1C=CC=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results