Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Furfurylpyrrole 98.0+%, TCI America™
SDP

Catalog No. F07005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07005G Supplier TCI America Supplier No. F07005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1438-94-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00010080
InChI Key BTBFUBUCCJKJOZ-UHFFFAOYSA-N
Synonym 1-furfurylpyrrole, n-furfurylpyrrole, 1-furan-2-ylmethyl-1h-pyrrole, 1-furfuryl-1h-pyrrole, 1h-pyrrole, 1-2-furanylmethyl, 1-2-furanylmethyl-1h-pyrrole, n-2-furfuryl pyrrole, n-furfuryl pyrrole, pyrrole, 1-furfuryl, 1-2-furfuryl pyrrole
PubChem CID 15037
IUPAC Name 1-[(furan-2-yl)methyl]-1H-pyrrole
SMILES C(N1C=CC=C1)C1=CC=CO1

Specifications

Boiling Point 79°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Furfurylpyrrole
RTECSNumber UX9631000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.