accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 14382-86-6

IUPAC Name: 2-(benzyloxy)-4,5-dimethoxybenzaldehyde
Synonyms: 2-(benzyloxy)-4,5-dimethoxybenzaldehyde
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: KTKIPTMZBXMHST-UHFFFAOYSA-N
SMILES: COC1=CC(OCC2=CC=CC=C2)=C(C=O)C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results