accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14384-71-5

IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)pyridine
Synonyms: 2-(2-methyl-1,3-thiazol-4-yl)pyridine
Molecular Weight (g/mol): 176.24
Molecular Formula: C9H8N2S
InChi Key: SSRDDRNVMGAKMJ-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results