Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-(2-pyridyl)thiazole, 97%, Thermo Scientific™

Catalog No. AAH5887809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5887809 10 g
AAH5887804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5887809 Supplier Thermo Scientific Chemicals Supplier No. H5887809

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14384-71-5
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD22666465
InChI Key SSRDDRNVMGAKMJ-UHFFFAOYSA-N
Synonym 2-methyl-4-2-pyridyl thiazole, 2-2-methyl-1,3-thiazol-4-yl pyridine, 2-methyl-4-pyridin-2-yl thiazole
PubChem CID 17845841
IUPAC Name 2-methyl-4-pyridin-2-yl-1,3-thiazole
SMILES CC1=NC(=CS1)C1=CC=CC=N1

Specifications

Melting Point 180°C to 185°C
Color Brown
Quantity 10 g
Beilstein 1565687
Formula Weight 176.24
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Methyl-4-(2-pyridyl)thiazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.