accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 143886-11-7

IUPAC Name: 9,9-dimethyl-N2,N7-bis(3-methylphenyl)-N2,N7-diphenyl-9H-fluorene-2,7-diamine
Synonyms: 9,9-dimethyl-N2,N7-bis(3-methylphenyl)-N2,N7-diphenylfluorene-2,7-diamine
Molecular Weight (g/mol): 556.75
Molecular Formula: C41H36N2
InChi Key: YUBXDAMWVRMLOG-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC(C)=C1)C2(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results