Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Dimethyl-2,7-bis[N-(m-tolyl)anilino]fluorene 98.0+%, TCI America™
SDP

Catalog No. D4920200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4920200MG 200 mg
D49201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4920200MG Supplier TCI America Supplier No. D4920200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143886-11-7
Molecular Formula C41H36N2
Molecular Weight (g/mol) 556.75
MDL Number MFCD11112144
InChI Key YUBXDAMWVRMLOG-UHFFFAOYSA-N
Synonym 9,9-Dimethyl-N,N′C-diphenyl-N,N′C-di(m-tolyl)fluorene-2,7-diamine, DMFL-TPD
PubChem CID 15404169
IUPAC Name 9,9-dimethyl-N2,N7-bis(3-methylphenyl)-N2,N7-diphenyl-9H-fluorene-2,7-diamine
SMILES CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC(C)=C1)C2(C)C

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 556.75
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Dimethyl-2,7-bis[N-(m-tolyl)anilino]fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.