accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNN

CAS RN 14389-12-9

IUPAC Name: 5-(pyridin-4-yl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(pyridin-4-yl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 146.13
Molecular Formula: C6H4N5
InChi Key: VSESUJHYHBRPFK-UHFFFAOYSA-N
SMILES: [N-]1N=NC(=N1)C1=CC=NC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results