Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals

Catalog No. AAB2074103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2074103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2074103 Supplier Thermo Scientific Chemicals Supplier No. B2074103
  • $81.00
    $68.65 / Each of 1

    Save $12.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14389-12-9 | Molecular Formula: C6H5N5 | Molecular Weight: 147.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14389-12-9
Molecular Formula C6H5N5
Molecular Weight (g/mol) 147.141
MDL Number MFCD00068115
InChI Key JMILUUVWLRKJFB-UHFFFAOYSA-N
Synonym 5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
PubChem CID 482249
IUPAC Name 4-(2H-tetrazol-5-yl)pyridine
SMILES C1=CN=CC=C1C2=NNN=N2

Specifications

Quantity 1 g
Formula Weight 147.14
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-(4-Pyridyl)-1H-tetrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.