accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2Br

CAS RN 1439-10-7

IUPAC Name: 5-bromopyrimidin-4-amine
Synonyms: 5-bromopyrimidin-4-amine
Molecular Weight (g/mol): 174
Molecular Formula: C4H4BrN3
InChi Key: IIFAONYUCDAVGA-UHFFFAOYSA-N
SMILES: NC1=NC=NC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results