accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 14392-69-9

IUPAC Name: 1-(4-hydroxyphenyl)nonan-1-one
Synonyms: 1-(4-hydroxyphenyl)nonan-1-one
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: JZBXYOOARNRUME-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)C1=CC=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results