accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144-39-8

IUPAC Name: 3,7-dimethylocta-1,6-dien-3-yl propanoate
Synonyms: linalyl propionate
Molecular Weight (g/mol): 210.32
Molecular Formula: C13H22O2
InChi Key: WAQIIHCCEMGYKP-UHFFFAOYNA-N
SMILES: CCC(=O)OC(C)(CCC=C(C)C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results