Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linalyl Propionate 97.0+%, TCI America™
SDP

Catalog No. L026525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L026525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L026525ML Supplier TCI America Supplier No. L026525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 144-39-8
Molecular Formula C13H22O2
Molecular Weight (g/mol) 210.317
MDL Number MFCD00048607
InChI Key WAQIIHCCEMGYKP-UHFFFAOYSA-N
Synonym Propionic Acid Linalyl Ester
PubChem CID 61098
IUPAC Name 3,7-dimethylocta-1,6-dien-3-yl propanoate
SMILES CCC(=O)OC(C)(CCC=C(C)C)C=C

Specifications

Color Colorless
Boiling Point 122°C
Quantity 25 mL
Formula Weight 210.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Linalyl Propionate
EINECSNumber (2)-0763
RTECSNumber RG5927903
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.