accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 14400-67-0

IUPAC Name: 2,5-dimethyl-2,3-dihydrofuran-3-one
Synonyms: 2,5-dimethyl-2H-furan-3-one
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: ASOSVCXGWPDUGN-UHFFFAOYNA-N
SMILES: CC1OC(C)=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results