Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3(2H)-furanone 98.0+%, TCI America™
SDP

Catalog No. D37955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D37955G Supplier TCI America Supplier No. D37955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 14400-67-0
Color Yellow
Boiling Point 72°C
Molecular Formula C6H8O2
Quantity 5 g
MDL Number MFCD00052571
UN Number 1993
Synonym 2,5-dimethyl-3 2h-furanone, 2,5-dimethylfuran-3 2h-one, 2,5-dimethyl-2,3-dihydrofuran-3-one, 3 2h-furanone, 2,5-dimethyl, 2,5-dimethyl-3 2h furanone, 2,5-dimethyl-2h-furan-3-one, 2,5-dimethyl-3-2h-furanone, 2h-furan-3-one, 2,5-dimethyl, mango furanone
InChI Key ASOSVCXGWPDUGN-UHFFFAOYSA-N
SMILES CC1C(=O)C=C(O1)C
IUPAC Name 2,5-dimethylfuran-3-one
Molecular Weight (g/mol) 112.128
PubChem CID 85730
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-3(2H)-furanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.