accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHONHNOONOOOH

CAS RN 14401-07-1

IUPAC Name: 2-[(2,4-dinitrophenyl)amino]-3-hydroxybutanoic acid
Synonyms: 2-[(2,4-dinitrophenyl)amino]-3-hydroxybutanoic acid
Molecular Weight (g/mol): 285.21
Molecular Formula: C10H11N3O7
InChi Key: PWOCOTZWYFGDMO-UHFFFAOYNA-N
SMILES: CC(O)C(NC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results