Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,4-Dinitrophenyl)-DL-threonine 99.0+%, TCI America™
SDP

Catalog No. D1039100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1039100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1039100MG Supplier TCI America Supplier No. D1039100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14401-07-1
Molecular Formula C10H11N3O7
Molecular Weight (g/mol) 285.212
MDL Number MFCD00070530
InChI Key PWOCOTZWYFGDMO-UHFFFAOYSA-N
Synonym N-Dnp-DL-threonine, Dnp-DL-Thr-OH
PubChem CID 262558
IUPAC Name 2-(2,4-dinitroanilino)-3-hydroxybutanoic acid
SMILES CC(C(C(=O)O)NC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-])O

Specifications

Melting Point 154°C
Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 285.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-DL-threonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.