accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 144069-67-0

OOHOON(S)CH3CH3H3C

CAS RN 144069-67-0

IUPAC Name: (2S)-1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indole-2-carboxylic acid
Synonyms: (2S)-1-(tert-butoxycarbonyl)-2,3-dihydroindole-2-carboxylic acid
Molecular Weight (g/mol): 263.29
Molecular Formula: C14H17NO4
InChi Key: QONNUMLEACJFME-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N1[C@@H](CC2=C1C=CC=C2)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)