accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(S)(R)OH(S)O(S)(S)HOOOOHNClBr

CAS RN 144110-43-0

IUPAC Name: (2S,3S,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: (2S,3S,4S,5R,6S)-6-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 421.6
Molecular Formula: C14H12BrClNO7
InChi Key: GAZVSOQUFLIBJQ-BYNIDDHOSA-M
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=C(Br)C=C23)O[C@@H]([C@H]1O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results