accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNH3COH

CAS RN 14426-20-1

IUPAC Name: hydrogen 2-(diethylamino)ethan-1-ol chloride
Synonyms: hydrogen DEAE chloride
Molecular Weight (g/mol): 153.65
Molecular Formula: C6H16ClNO
InChi Key: DBQUKJMNGUJRFI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)CCO
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)