CAS RN 144335-37-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ FGF/PDGF/VEGF RTK Inhibitor
The FGF/PDGF/VEGF RTK Inhibitor, also referenced under CAS 144335-37-5, controls the biological activity of FGF/PDGF/VEGF RTK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.