CAS RN 1443437-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MICAL-2 Inhibitor II, CCG-203971
A cell-permeable, restricted form of CCG-1423 with enhanced potency (IC50 = 4.2 μM vs 16.6 μM for PC-3 cell migration), but reduced off-target toxicity.