Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MICAL-2 Inhibitor II, CCG-203971

Catalog No. 5309850001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-098-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-098-50001 Supplier MilliporeSigma™ Supplier No. 5309850001
Add to Cart
Add to Cart

A cell-permeable, restricted form of CCG-1423 with enhanced potency (IC50 = 4.2 μM vs 16.6 μM for PC-3 cell migration), but reduced off-target toxicity.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
Target Proteins MICAL-2
CAS 1443437-74-8
Molecular Formula C23H21ClN2O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.