accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14436-32-9

IUPAC Name: dec-9-enoic acid
Synonyms: 9-decenoic acid delta(9)-decenoic acid caproleic acid 9-decylenic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: KHAVLLBUVKBTBG-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results