accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiBFFFFFFCH3OOOH3C

CAS RN 1443685-69-5

IUPAC Name: lithium(1+) dimethyl carbonate trifluoro(trifluoromethyl)boranuide
Synonyms: lithium(1+) dimethyl carbonate trifluoro(trifluoromethyl)boranuide
Molecular Weight (g/mol): 233.83
Molecular Formula: C4H6BF6LiO3
InChi Key: ZGAWBZORCLPFJC-UHFFFAOYSA-N
SMILES: [Li+].COC(=O)OC.F[B-](F)(F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results