accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNO

CAS RN 1444-94-6

IUPAC Name: octahydro-1H-isoindole-1,3-dione
Synonyms: hexahydro-2H-isoindole-1,3-dione
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: WLDMPODMCFGWAA-UHFFFAOYNA-N
SMILES: O=C1NC(=O)C2CCCCC12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results