Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexahydrophthalimide 98.0+%, TCI America™
SDP

Catalog No. H136325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H136325G 25 g
H1363250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H136325G Supplier TCI America Supplier No. H136325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1444-94-6
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00035737
InChI Key WLDMPODMCFGWAA-UHFFFAOYSA-N
Synonym 1,2-Cyclohexanedicarboximide
PubChem CID 73559
IUPAC Name 3a,4,5,6,7,7a-hexahydroisoindole-1,3-dione
SMILES C1CCC2C(C1)C(=O)NC2=O

Specifications

Melting Point 137°C
Color White
Quantity 25 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexahydrophthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.