accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNO

CAS RN 1445-69-8

IUPAC Name: 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Synonyms: 4a,8a-dihydrophthalazine-1,4-dione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: JNQUJPBFSQETPT-UHFFFAOYNA-N
SMILES: O=C1N=NC(=O)C2C=CC=CC12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)