Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHTHALIC HYDRAZIDE , MP Biomedicals

Catalog No. ICN216707 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216707 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216707 Supplier MP Biomedicals, Inc Supplier No. 0521670780

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00006888
InChI Key JNQUJPBFSQETPT-UHFFFAOYNA-N
Synonym 2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazine
PubChem CID 219401
IUPAC Name 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
SMILES O=C1N=NC(=O)C2C=CC=CC12

Specifications

Quantity 100 g
Formula Weight 162.15
Chemical Name or Material 1, 4-Phthalazinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.