accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1445-73-4

IUPAC Name: 1-methylpiperidin-4-one
Synonyms: 1-methylpiperidin-4-one
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: HUUPVABNAQUEJW-UHFFFAOYSA-N
SMILES: CN1CCC(=O)CC1
Percent Purity 
  • (3)
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (5)