accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14456-34-9

IUPAC Name: dichloro(oxo)hafnium octahydrate
Synonyms: dichloro(oxo)hafnium octahydrate
Molecular Weight (g/mol): 409.51
Molecular Formula: Cl2H16HfO9
InChi Key: FXQGUVLIMYOGDZ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)