Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr <1.5%

Catalog No. AA1183336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1183336 500 g
AA1183314 25 g
AA1183322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1183336 Supplier Thermo Scientific Chemicals Supplier No. 01183336
Add to Cart

missing translation for 'validMainframeMsg'

For Synthesis of Hafnium (IV) Oxide | CAS: 14456-34-9 | Cl2H16HfO9 | 409.51 g/mol

Hafnium dichloride oxide octahydrate is used in the synthesis of hafnium(IV) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-34-9
Molecular Formula Cl2H16HfO9
Molecular Weight (g/mol) 409.51
MDL Number MFCD00150417
InChI Key FXQGUVLIMYOGDZ-UHFFFAOYSA-L
Synonym hafnium iv dichloride oxide octahydrate 98+%-hf zr
PubChem CID 131675691
IUPAC Name hafnium;dichloride;nonahydrate
SMILES O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water.
Formula Weight 409.52 (265.40 Anhydrous)
Odor Pungent
Sensitivity Hygroscopic
Density 2.2
Chemical Name or Material Hafnium dichloride oxide octahydrate
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packing Group: III
EINECSNumber 237-345-3
RTECSNumber MG4800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.