accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14456-48-5

BrBrBrDy

CAS RN 14456-48-5

IUPAC Name: dysprosium(3+) tribromide
Synonyms: dysprosium(3+) tribromide
Molecular Weight (g/mol): 402.21
Molecular Formula: Br3Dy
InChi Key: GBLDKMKYYYAAKD-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Dy+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results