accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrFFF

CAS RN 144584-67-8

IUPAC Name: 1-bromo-2-fluoro-3-(trifluoromethyl)benzene
Synonyms: 1-bromo-2-fluoro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 243
Molecular Formula: C7H3BrF4
InChi Key: VPXCYIIXCVJZKZ-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results