Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. B33895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33895G 5 g
B33891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33895G Supplier TCI America Supplier No. B33895G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 144584-67-8
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD00070812
InChI Key VPXCYIIXCVJZKZ-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzotrifluoride, 1-bromo-2-fluoro-3-trifluoromethyl benzene, 2-fluoro-3-trifluoromethyl bromobenzene, 1-bromo-2-fluoro-3-trifluoromethyl-benzene, 3-bromo-2-fluoro-1-trifluoromethyl benzene, benzene, 1-bromo-2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethylbromobenzene, pubchem4840, intermediates-zcf02110, 3-bromo-alpha,alpha,alpha,2-tetrafluorotoluene
PubChem CID 2736320
IUPAC Name 1-bromo-2-fluoro-3-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)Br)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 169°C
Quantity 5 g
Formula Weight 243.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-fluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.