accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNNHNNHOOCH3H3COH

CAS RN 144689-24-7

IUPAC Name: 4-(2-hydroxypropan-2-yl)-2-propyl-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-imidazole-5-carboxylic acid
Synonyms: 5-(2-hydroxypropan-2-yl)-2-propyl-3-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}imidazole-4-carboxylic acid
Molecular Weight (g/mol): 446.51
Molecular Formula: C24H26N6O3
InChi Key: VTRAEEWXHOVJFV-UHFFFAOYSA-N
SMILES: CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(O)=O)C(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results