Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olmesartan, Tocris Bioscience™
SDP

Catalog No. 461650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-165-0 Supplier R&D Systems Supplier No. 4616/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent AT1 antagonist; metabolite of Olmesartan medoxomil (Cat. No. 4620)

Potent angiotensin II type I (AT1) receptor antagonist (IC50 = 6.7 nM at hAT1 receptors). Improves cognitive function in a mouse model of hypertension- and hyperlipidemia-associated cognitive decline; ameliorates arterial stiffness in a rat model of chronic renal failure.

Chemical Identifiers

CAS 144689-24-7
Molecular Formula C24H26N6O3
Molecular Weight (g/mol) 446.51
MDL Number MFCD00914967
InChI Key VTRAEEWXHOVJFV-UHFFFAOYSA-N
Synonym olmesartan, olmesartan acid, votum, olme sartan, unii-8w1iqp3u10, chembl1516, 4-2-hydroxypropan-2-yl-2-propyl-1-4-2-1h-1,2,3,4-tetrazol-5-yl phenyl phenyl methyl-1h-imidazole-5-carboxylic acid, 1-2'-1h-tetrazol-5-yl-1,1'-biphenyl-4-yl methyl-4-2-hydroxypropan-2-yl-2-propyl-1h-imidazole-5-carboxylic acid, 1h-imidazole-5-carboxylic acid, 4-1-hydroxy-1-methylethyl-2-propyl-1-2'-1h-tetrazol-5-yl 1,1'-biphenyl-4-yl methyl, 4-1-hydroxy-1-methylethyl-2-propyl-1-2'-2h-tetrazol-5-yl 1,1'-biphenyl-4-yl methyl-1h-imidazole-5-carboxylic acid
PubChem CID 158781
ChEBI CHEBI:48416
IUPAC Name 4-(2-hydroxypropan-2-yl)-2-propyl-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-imidazole-5-carboxylic acid
SMILES CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(O)=O)C(C)(C)O

Specifications

Quantity 50 mg
Formula Weight 446.5
Percent Purity >99%
Chemical Name or Material Olmesartan
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.