accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNH3CH3COHOOCH3

CAS RN 144689-93-0

IUPAC Name: ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-4-carboxylate
Synonyms: ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-4-carboxylate
Molecular Weight (g/mol): 240.3
Molecular Formula: C12H20N2O3
InChi Key: KZBJJAFGNMRRHN-UHFFFAOYSA-N
SMILES: CCCC1=NC(C(=O)OCC)=C(N1)C(C)(C)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results