Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylate 96.0+%, TCI America™
SDP

Catalog No. E097625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E097625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E097625G Supplier TCI America Supplier No. E097625G

missing translation for 'validMainframeMsg'

Specifications

CAS 144689-93-0
Melting Point 104°C
Color Yellow
Molecular Formula C12H20N2O3
Quantity 25 g
MDL Number MFCD07787565
Synonym 4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylic Acid Ethyl Ester
InChI Key KZBJJAFGNMRRHN-UHFFFAOYSA-N
SMILES CCCC1=NC(=C(N1)C(C)(C)O)C(=O)OCC
IUPAC Name ethyl 5-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-4-carboxylate
Molecular Weight (g/mol) 240.303
PubChem CID 10562026
Formula Weight 240.30
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-(2-Hydroxy-2-propyl)-2-propyl-1H-imidazole-5-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.