accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONHON

CAS RN 1448-98-2

IUPAC Name: (2-cyanoacetyl)urea
Synonyms: 2-cyanoacetylurea
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: QJGRPCPCQQPZLZ-UHFFFAOYSA-N
SMILES: NC(=O)NC(=O)CC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results