Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanoacetylurea 98.0+%, TCI America™
SDP

Catalog No. C110125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110125G 25 g
C1101250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C110125G Supplier TCI America Supplier No. C110125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1448-98-2
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00007947
InChI Key QJGRPCPCQQPZLZ-UHFFFAOYSA-N
Synonym cyanoacetylurea, 2-cyanoacetylurea, acetamide, n-aminocarbonyl-2-cyano, cyanoacetyl urea, urea, cyanoacetyl, 2-cyanoacetyl urea, amino-n-2-cyanoacetyl amide, n-2-cyanoacetyl urea, n-cyanoacetyl urea
PubChem CID 74055
IUPAC Name N-carbamoyl-2-cyanoacetamide
SMILES C(C#N)C(=O)NC(=O)N

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 127.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyanoacetylurea
EINECSNumber (2)-3324
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.