accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSFFFFFOOH

CAS RN 1448319-14-9

IUPAC Name: 2-methoxy-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 2-methoxy-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 278.19
Molecular Formula: C8H7F5O3S
InChi Key: STKWSTMSPXNSCO-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results