Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-(pentafluorothio)benzoic acid, 97%, Thermo Scientific™

Catalog No. AAH34072MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34072MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH34072MD Supplier Thermo Scientific Chemicals Supplier No. H34072MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1448319-14-9
Molecular Formula C8H7F5O3S
Quantity 250mg
MDL Number MFCD16652486
Synonym 2-methoxy-5-pentafluorosulfur benzoic acid, 2-methoxy-5-pentafluoro-??-sulfanyl benzoic acid, 2-methoxy-5-pentafluorothio benzoic acid, 2-methoxy-5-pentafluoro-lambda6-sulphanyl benzoic acid
InChI Key STKWSTMSPXNSCO-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)C(=O)O
IUPAC Name 2-methoxy-5-(pentafluoro-$l^{6}-sulfanyl)benzoic acid
Molecular Weight (g/mol) 278.193
PubChem CID 66523609
Formula Weight 278.19
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(pentafluorothio)benzoic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.