accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNH2ONHOHO

CAS RN 14486-03-4

IUPAC Name: 2-[2-amino-4-(methylsulfanyl)butanamido]acetic acid
Synonyms: [2-amino-4-(methylsulfanyl)butanamido]acetic acid
Molecular Weight (g/mol): 206.26
Molecular Formula: C7H14N2O3S
InChi Key: QXOHLNCNYLGICT-UHFFFAOYNA-N
SMILES: CSCCC(N)C(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results